Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Task6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275612.1 | NP_001262541.1 | 7 - 8 | 3110 | 93 | |
NM_142043.3 | NP_650300.2 | 7 - 8 | 2062 | 93 | ||
GenBank mRNA | AJ920330.1 | - | 6 - 7 | 950 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |