Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260184.1 | NP_001247113.1 | 6 - 7 | 925 | 60 | |
NM_001260185.1 | NP_001247114.1 | 6 - 7 | 959 | 60 | ||
NM_142166.2 | NP_650423.1 | 6 - 7 | 959 | 60 | ||
GenBank mRNA | AY071198.1 | - | 6 - 7 | 959 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |