Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202307.2 | NP_001189236.1 | 4 - 5 | 812 | 58 | |
NM_001202308.2 | NP_001189237.1 | 4 - 5 | 797 | 58 | ||
NM_142315.4 | NP_650572.2 | 4 - 5 | 753 | 58 | ||
NM_169727.2 | NP_732167.1 | 3 - 4 | 891 | 58 | ||
GenBank mRNA | AY089543.1 | - | 4 - 5 | 721 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |