Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275750.1 | NP_001262679.1 | 3 - 4 | 2066 | 64 | |
NM_142388.3 | NP_650645.1 | 3 - 4 | 2156 | 64 | ||
GenBank mRNA | AY051768.1 | - | 3 - 4 | 2156 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |