Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104379.2 | NP_001097849.1 | 10 - 11 | 1935 | 61 | |
NM_169925.3 | NP_732555.1 | 10 - 11 | 1761 | 61 | ||
NM_169927.3 | NP_732557.2 | 11 - 12 | 2022 | 61 | ||
NM_169928.3 | NP_732558.2 | 10 - 11 | 2461 | 61 | ||
NM_169929.3 | NP_732559.1 | 10 - 11 | 2012 | 61 | ||
GenBank mRNA | AY058477.1 | - | 10 - 11 | 2310 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |