Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043289.2 | NP_001036754.1 | 5 - 6 | 5984 | 59 | |
NM_001043290.2 | NP_001036755.2 | 5 - 6 | 5981 | 59 | ||
NM_001260359.1 | NP_001247288.1 | 4 - 5 | 5918 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |