Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp6a18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300633.1 | NP_001287562.1 | 1 - 2 | 1135 | 77 | |
NM_143306.3 | NP_651563.1 | 1 - 2 | 1135 | 77 | ||
GenBank mRNA | AY071195.1 | - | 1 - 2 | 1133 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |