Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WASp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276116.1 | NP_001263045.1 | 3 - 4 | 550 | 65 | |
NM_143380.4 | NP_651637.1 | 3 - 4 | 610 | 65 | ||
NM_170378.2 | NP_733257.1 | 3 - 4 | 498 | 65 | ||
NM_176578.2 | NP_788755.1 | 2 - 3 | 316 | 65 | ||
GenBank mRNA | AY070576.1 | - | 3 - 4 | 621 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |