Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNF4Agamma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170200.2 | NP_001163671.1 | 3 - 4 | 2006 | 89 | |
NM_001170201.2 | NP_001163672.1 | 3 - 4 | 1888 | 89 | ||
NM_169947.3 | NP_732598.1 | 3 - 4 | 2065 | 89 | ||
GenBank mRNA | AY070541.1 | - | 3 - 4 | 2042 | 89 | |
AY166752.1 | - | 3 - 4 | 2049 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |