Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp96F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276047.2 | NP_001262976.1 | 3 - 4 | 778 | 72 | |
NM_001276048.2 | NP_001262977.1 | 3 - 4 | 778 | 72 | ||
NM_079776.3 | NP_524500.1 | 3 - 4 | 778 | 72 | ||
GenBank mRNA | AF160941.1 | - | 3 - 4 | 725 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |