Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ser | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300629.1 | NP_001287558.1 | 10 - 11 | 2908 | 149 | |
NM_079803.3 | NP_524527.3 | 10 - 11 | 2636 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |