Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260472.2 | NP_001247401.1 | 3 - 4 | 1397 | 57 | |
NM_079875.3 | NP_524614.2 | 5 - 6 | 1730 | 57 | ||
GenBank mRNA | AY058519.1 | - | 3 - 4 | 1360 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |