Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Axn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144654.3 | NP_001138126.1 | 5 - 6 | 1791 | 75 | |
NM_001276168.1 | NP_001263097.1 | 5 - 6 | 1791 | 75 | ||
NM_001276169.1 | NP_001263098.1 | 3 - 4 | 738 | 75 | ||
NM_170457.3 | NP_733336.1 | 5 - 6 | 1791 | 75 | ||
NM_170458.2 | NP_733337.1 | 3 - 4 | 738 | 75 | ||
NM_170459.3 | NP_733338.1 | 5 - 6 | 1791 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |