Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PH4alphaNE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170499.2 | NP_733378.1 | 2 - 3 | 352 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |