Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104512.4 | NP_001097982.1 | 2 - 3 | 444 | 74 | |
NM_001170305.3 | NP_001163776.1 | 2 - 3 | 759 | 74 | ||
NM_001202391.2 | NP_001189320.1 | 2 - 3 | 421 | 74 | ||
NM_001260442.2 | NP_001247371.1 | 2 - 3 | 438 | 74 | ||
NM_001260443.2 | NP_001247372.1 | 2 - 3 | 552 | 74 | ||
NM_001276182.1 | NP_001263111.1 | 1 - 2 | 424 | 74 | ||
NM_001276183.1 | NP_001263112.1 | 1 - 2 | 424 | 74 | ||
NM_001382033.1 | NP_001369033.1 | 2 - 3 | 444 | 74 | ||
NM_170503.4 | NP_733382.1 | 1 - 2 | 665 | 74 | ||
NM_170504.4 | NP_733383.1 | 2 - 3 | 444 | 74 | ||
NM_170505.4 | NP_733384.1 | 2 - 3 | 529 | 74 | ||
GenBank mRNA | BT133473.1 | - | 2 - 3 | 437 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |