Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078842.5 | NP_523566.4 | 2 - 3 | 2012 | 138 | |
NM_205990.2 | NP_995712.1 | 2 - 3 | 2009 | 138 | ||
GenBank mRNA | BT001713.1 | - | 2 - 3 | 2007 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |