Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201594.2 | NP_001188523.1 | 2 | 736 | 56 | |
NM_080298.4 | NP_525037.1 | 2 | 828 | 56 | ||
GenBank mRNA | BT014915.1 | - | 2 | 803 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |