Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: achi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299423.1 | NP_001286352.1 | 9 | 2111 | 131 | |
NM_165911.2 | NP_725182.1 | 8 | 1956 | 131 | ||
NM_165912.2 | NP_725183.1 | 9 | 2343 | 131 | ||
GenBank mRNA | BT003510.1 | - | 8 | 1946 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |