Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CkIIalpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043159.3 | NP_001036624.1 | 3 | 1252 | 115 | |
NM_001300222.1 | NP_001287151.1 | 2 | 1152 | 115 | ||
NM_001300223.1 | NP_001287152.1 | 2 | 1100 | 115 | ||
NM_001300224.1 | NP_001287153.1 | 4 | 1724 | 115 | ||
NM_080179.5 | NP_524918.1 | 3 | 1232 | 115 | ||
NM_168985.2 | NP_730774.1 | 3 | 1207 | 115 | ||
NM_168986.2 | NP_730775.1 | 2 | 1094 | 115 | ||
GenBank mRNA | AY069553.1 | - | 3 | 1148 | 115 | |
M16534.1 | - | 3 | 1138 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |