Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipped-B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042986.3 | NP_001036451.1 | 14 - 15 | 4108 | 141 | |
NM_001042987.3 | NP_001036452.1 | 14 - 15 | 4136 | 141 | ||
NM_001169581.2 | NP_001163052.1 | 15 - 16 | 4029 | 141 | ||
NM_001169582.2 | NP_001163053.1 | 15 - 16 | 4064 | 141 | ||
NM_001169583.2 | NP_001163054.1 | 15 - 16 | 4036 | 141 | ||
NM_001273774.1 | NP_001260703.1 | 13 - 14 | 3776 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |