Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cep43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002125.1 | NP_001002125.1 | 15 - 16 | 1298 | 152 | |
XM_021468391.1 | XP_021324066.1 | 17 - 18 | 1470 | 152 | ||
GenBank mRNA | BC071450.1 | - | 15 - 16 | 1298 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |