Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: supt7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013292.2 | NP_001013310.1 | 2 - 3 | 589 | 66 | |
GenBank mRNA | BC090267.1 | - | 2 - 3 | 593 | 66 | |
BC129289.1 | - | 2 - 3 | 565 | 66 | ||
BC165795.1 | - | 1 - 2 | 461 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |