Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp2caa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017886.1 | NP_001017886.1 | 4 - 5 | 933 | 65 | |
GenBank mRNA | BC092961.1 | - | 4 - 5 | 933 | 65 | |
BC155238.1 | - | 4 - 5 | 863 | 65 | ||
BC165383.1 | - | 4 - 5 | 616 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |