Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tyrobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100103.2 | NP_001093573.2 | 3 - 4 | 293 | 58 | |
XM_009292219.1 | XP_009290494.1 | 3 - 4 | 1266 | 58 | ||
GenBank mRNA | EF158446.1 | - | 3 - 4 | 293 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Danio rerio Probe spans exons |