Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alcama | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131000.1 | NP_571075.1 | 3 - 4 | 538 | 111 | |
XM_005172635.4 | XP_005172692.1 | 3 - 4 | 564 | 111 | ||
GenBank mRNA | BC050482.1 | - | 3 - 4 | 474 | 111 | |
BC071276.1 | - | 3 - 4 | 539 | 111 | ||
CU458879.1 | - | 3 - 4 | 379 | 111 | ||
CU458962.1 | - | 3 - 4 | 485 | 111 | ||
L25273.1 | - | 3 - 4 | 538 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |