Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131051.1 | NP_571126.1 | 3 - 4 | 1188 | 90 | |
XM_017359202.2 | XP_017214691.1 | 1 - 2 | 131 | 90 | ||
GenBank mRNA | AF136946.1 | - | 3 - 4 | 741 | 90 | |
AF179405.1 | - | 3 - 4 | 1188 | 90 | ||
BC051603.1 | - | 3 - 4 | 1184 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |