Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csnk2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131252.1 | NP_571327.1 | 10 - 11 | 897 | 69 | |
XM_005158170.3 | XP_005158227.1 | 10 - 11 | 945 | 69 | ||
GenBank mRNA | BC044403.1 | - | 10 - 11 | 897 | 69 | |
BC067595.1 | - | 9 - 10 | 814 | 69 | ||
BC165881.1 | - | 7 - 8 | 655 | 69 | ||
S76875.1 | - | 6 - 7 | 437 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |