Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snrnp40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200322.1 | NP_956616.1 | 9 - 10 | 1097 | 73 | |
GenBank mRNA | BC051783.1 | - | 9 - 10 | 1097 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |