Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: akr1b1.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002048.1 | NP_001002048.1 | 3 - 4 | 419 | 84 | |
GenBank mRNA | BC071313.1 | - | 3 - 4 | 419 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |