Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045044.1 | NP_001038509.1 | 12 - 13 | 1269 | 75 | |
XM_005161685.4 | XP_005161742.1 | 12 - 13 | 1331 | 75 | ||
XM_009296204.3 | XP_009294479.1 | 12 - 13 | 1338 | 75 | ||
XM_021469341.1 | XP_021325016.1 | 6 - 7 | 3635 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Danio rerio Probe spans exons |