Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: loxl5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045856.2 | NP_001039321.2 | 1 - 2 | 848 | 96 | |
XM_021469354.1 | XP_021325029.1 | 1 - 2 | 952 | 96 | ||
GenBank mRNA | EF030485.1 | - | 1 - 2 | 848 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Danio rerio Probe spans exons |