Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: il15ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083584.2 | NP_001077053.1 | 8 - 9 | 778 | 87 | |
NM_001136253.2 | NP_001129725.2 | 6 - 7 | 469 | 87 | ||
NM_001359926.1 | NP_001346855.1 | 7 - 8 | 760 | 87 | ||
NM_001359927.1 | NP_001346856.1 | 7 - 8 | 556 | 87 | ||
NM_001359928.1 | NP_001346857.1 | 6 - 7 | 538 | 87 | ||
NM_001359929.1 | NP_001346858.1 | 6 - 7 | 505 | 87 | ||
NM_001359930.1 | NP_001346859.1 | 5 - 6 | 487 | 87 | ||
NM_001359931.1 | NP_001346860.1 | 5 - 6 | 451 | 87 | ||
NM_001359932.1 | NP_001346861.1 | 5 - 6 | 418 | 87 | ||
NM_001359933.1 | NP_001346862.1 | 4 - 5 | 400 | 87 | ||
NR_153373.1 | 4 - 5 | 256 | 87 | |||
XM_005164796.3 | XP_005164853.1 | 7 - 8 | 804 | 87 | ||
GenBank mRNA | BC163782.1 | - | 6 - 7 | 464 | 87 | |
EF143578.1 | - | 8 - 9 | 778 | 87 | ||
EF165721.1 | - | 7 - 8 | 760 | 87 | ||
EF165722.1 | - | 7 - 8 | 556 | 87 | ||
EF165723.1 | - | 6 - 7 | 538 | 87 | ||
EF165724.1 | - | 6 - 7 | 505 | 87 | ||
EF165725.1 | - | 5 - 6 | 487 | 87 | ||
EF165726.1 | - | 6 - 7 | 469 | 87 | ||
EF165727.1 | - | 5 - 6 | 451 | 87 | ||
EF165728.1 | - | 5 - 6 | 441 | 87 | ||
EF165729.1 | - | 5 - 6 | 418 | 87 | ||
EF165730.1 | - | 4 - 5 | 400 | 87 | ||
EF165731.1 | - | 4 - 5 | 256 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |