Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inhbb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131068.2 | NP_571143.2 | 1 - 2 | 851 | 67 | |
GenBank mRNA | BC162140.1 | - | 1 - 2 | 822 | 67 | |
BC162143.1 | - | 1 - 2 | 822 | 67 | ||
X76051.1 | - | 858 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |