Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc46a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200285.1 | NP_956579.1 | 5 - 6 | 1477 | 74 | |
GenBank mRNA | BC049421.1 | - | 5 - 6 | 1477 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |