Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gmppab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200497.1 | NP_956791.1 | 5 - 6 | 728 | 73 | |
GenBank mRNA | BC055506.1 | - | 5 - 6 | 728 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |