Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hnrnpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201099.1 | NP_957393.1 | 5 - 6 | 649 | 86 | |
XM_021476043.1 | XP_021331718.1 | 5 - 6 | 671 | 86 | ||
GenBank mRNA | BC045336.1 | - | 5 - 6 | 649 | 86 | |
BC164693.1 | - | 5 - 6 | 661 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |