Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: seta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201475.1 | NP_958883.1 | 5 - 6 | 692 | 85 | |
GenBank mRNA | BC046082.1 | - | 5 - 6 | 692 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |