Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prpf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212655.2 | NP_997820.2 | 18 - 19 | 2580 | 65 | |
GenBank mRNA | BC056710.1 | - | 18 - 19 | 2556 | 65 | |
BC066556.1 | - | 18 - 19 | 2544 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |