Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arpc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213156.1 | NP_998321.1 | 8 - 9 | 1099 | 73 | |
GenBank mRNA | BC053229.1 | - | 8 - 9 | 1099 | 73 | |
BC164463.1 | - | 7 - 8 | 1017 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |