Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arhgdia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213461.1 | NP_998626.1 | 3 - 4 | 413 | 135 | |
GenBank mRNA | BC047172.1 | - | 3 - 4 | 413 | 135 | |
BC063968.1 | - | 3 - 4 | 384 | 135 | ||
BC164027.1 | - | 2 - 3 | 312 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |