Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: uqcrc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001589.1 | NP_001001589.1 | 5 - 6 | 449 | 88 | |
GenBank mRNA | BC054137.1 | - | 5 - 6 | 427 | 88 | |
BC071551.1 | - | 5 - 6 | 449 | 88 | ||
BC097011.1 | - | 5 - 6 | 408 | 88 | ||
BC152592.1 | - | 5 - 6 | 427 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |