Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbm42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003850.1 | NP_001003850.1 | 6 - 7 | 668 | 93 | |
GenBank mRNA | AY648798.1 | - | 6 - 7 | 668 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |