Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005981.1 | NP_001005981.1 | 3 - 4 | 438 | 82 | |
GenBank mRNA | BC083429.1 | - | 3 - 4 | 438 | 82 | |
BC163998.1 | - | 3 - 4 | 421 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |