Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nipsnap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131034.1 | NP_571109.1 | 813 | 83 | ||
GenBank mRNA | AJ249797.1 | - | 803 | 83 | ||
BC081660.1 | - | 813 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio |