Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: otpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131100.1 | NP_571175.1 | 2 - 3 | 547 | 123 | |
XM_005165505.3 | XP_005165562.1 | 2 - 3 | 610 | 123 | ||
GenBank mRNA | AF071496.2 | - | 2 - 3 | 547 | 123 | |
BC076366.1 | - | 2 - 3 | 558 | 123 | ||
BC164107.1 | - | 2 - 3 | 466 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |