Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tomm40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199614.1 | NP_955908.1 | 8 - 9 | 810 | 69 | |
XM_005158091.4 | XP_005158148.1 | 8 - 9 | 1218 | 69 | ||
XM_017351318.2 | XP_017206807.1 | 8 - 9 | 1007 | 69 | ||
GenBank mRNA | BC053295.1 | - | 8 - 9 | 810 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |