Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: her8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199624.3 | NP_955918.3 | 2 - 3 | 253 | 85 | |
XM_005172256.4 | XP_005172313.1 | 2 - 3 | 314 | 85 | ||
GenBank mRNA | AY007990.1 | - | 2 - 3 | 207 | 85 | |
AY007991.1 | - | 2 - 3 | 251 | 85 | ||
BC068368.1 | - | 2 - 3 | 226 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |