Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myoz1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205678.1 | NP_991241.1 | 3 - 4 | 346 | 68 | |
GenBank mRNA | BC065451.1 | - | 3 - 4 | 346 | 68 | |
BC155186.1 | - | 3 - 4 | 344 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |