Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lsm14ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214772.2 | NP_999937.2 | 4 - 5 | 772 | 65 | |
XM_005158973.3 | XP_005159030.1 | 5 - 6 | 871 | 65 | ||
XM_005158974.3 | XP_005159031.1 | 5 - 6 | 871 | 65 | ||
GenBank mRNA | AY577000.1 | - | 4 - 5 | 749 | 65 | |
BC065685.1 | - | 4 - 5 | 737 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |