Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adipor1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001327754.1 | NP_001314683.1 | 4 - 5 | 592 | 80 | |
GenBank mRNA | BC076349.1 | - | 4 - 5 | 80 | ||
BC165131.1 | - | 2 - 3 | 80 | |||
EH448747.1 | - | 4 - 5 | 518 | 80 | ||
EH505010.1 | - | 4 - 5 | 534 | 80 | ||
EH586253.1 | - | 4 - 5 | 80 | |||
EU139312.1 | - | 2 - 3 | 139 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |